missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Sulfate, Pentahydrate, Powder, USP, 98.5-100.5%, Spectrum™ Chemical
CU110, 7758-99-8, CuSO4·5H2O
$371.07 - $11488.30
Chemical Identifiers
| CAS | 7758-99-8 |
|---|---|
| Molecular Formula | CuH10O9S |
| Molecular Weight (g/mol) | 249.68 |
| MDL Number | MFCD00149681 |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| IUPAC Name | copper(2+) pentahydrate sulfate |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601925
|
Spectrum Chemical Mfg Cor
CU110125GM |
125 g | Poly Bottle |
Each for $371.07
|
|
||||
|
18601926
|
Spectrum Chemical Mfg Cor
CU110500GM |
500 g | Poly Bottle |
Each for $819.67
|
|
||||
|
18601927
|
Spectrum Chemical Mfg Cor
CU11025KG |
2.5 kg | Poly Bottle |
Each for $2,375.40
|
|
||||
|
18601928
|
Spectrum Chemical Mfg Cor
CU11012KG |
12 kg | Poly Pail |
Each for $5,236.63
|
|
||||
|
18601929
|
Spectrum Chemical Mfg Cor
CU11045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Cupric Sulfate, Pentahydrate, Powder, USP is used as a mineral supplement and in topical antifungal preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7758-99-8 | |
| 249.68 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| CuH10O9S | |
| MFCD00149681 | |
| copper(2+) pentahydrate sulfate |
Specifications
| 7758-99-8 | |
| 125 g | |
| MFCD00149681 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| copper(2+) pentahydrate sulfate | |
| 98.5 to 100.5% | |
| 33.0-36.5% |
| 100% | |
| CuH10O9S | |
| UN3077 | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
| 249.68 | |
| USP | |
| Poly Bottle |