missing translation for 'onlineSavingsMsg'
Learn More

Cupric Sulfate, Pentahydrate, Powder, USP, 98.5-100.5%, Spectrum™ Chemical

Catalog No. 18601929
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601929 45 kg Fiber Drum
18601925 125 g Poly Bottle
18601926 500 g Poly Bottle
18601927 2.5 kg Poly Bottle
18601928 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601929 Supplier Spectrum Chemical Mfg Cor Supplier No. CU11045KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CU110, 7758-99-8, CuSO4·5H2O

Spectrum™ Chemical Cupric Sulfate, Pentahydrate, Powder, USP is used as a mineral supplement and in topical antifungal preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 45 kg
UN Number UN3077
Percent Purity 98.5 to 100.5%
Grade USP
Loss on Drying 33.0-36.5%
Packaging Fiber Drum