missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Sulfate, Pentahydrate, Powder, USP, 98.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Cupric Sulfate, Pentahydrate, Powder, USP is used as a mineral supplement and in topical antifungal preparations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 125 g |
| UN Number | UN3077 |
| Percent Purity | 98.5 to 100.5% |
| Grade | USP |
| Loss on Drying | 33.0-36.5% |
| Packaging | Poly Bottle |