missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cumylamine 98.0+%, TCI America™
$272.16 - $814.59
Chemical Identifiers
| CAS | 585-32-0 |
|---|---|
| Molecular Formula | C9H13N |
| Molecular Weight (g/mol) | 135.21 |
| MDL Number | MFCD00134680 |
| InChI Key | KDFDOINBXBEOLZ-UHFFFAOYSA-N |
| Synonym | cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine |
| PubChem CID | 68509 |
| IUPAC Name | 2-phenylpropan-2-amine |
| SMILES | CC(C)(N)C1=CC=CC=C1 |
Chemical Identifiers
| 585-32-0 | |
| 135.21 | |
| KDFDOINBXBEOLZ-UHFFFAOYSA-N | |
| 68509 | |
| CC(C)(N)C1=CC=CC=C1 |
| C9H13N | |
| MFCD00134680 | |
| cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine | |
| 2-phenylpropan-2-amine |
Specifications
| 585-32-0 | |
| 197°C | |
| MFCD00134680 | |
| cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine | |
| CC(C)(N)C1=CC=CC=C1 | |
| 135.21 | |
| 135.21 | |
| Liquid |
| Yellow | |
| C9H13N | |
| 5 mL | |
| KDFDOINBXBEOLZ-UHFFFAOYSA-N | |
| 2-phenylpropan-2-amine | |
| 68509 | |
| ≥98.0% (GC,T) | |
| Cumylamine |
Safety and Handling
EINECSNumber : (3)-0332&(3)-2693
TSCA : No