accessibility menu, dialog, popup

UserName

CH3H3CNH2

CAS RN 585-32-0

IUPAC Name: 2-phenylpropan-2-amine
Synonyms: dimethyl-aminophenylmethane
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: KDFDOINBXBEOLZ-UHFFFAOYSA-N
SMILES: CC(C)(N)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)