missing translation for 'onlineSavingsMsg'
Learn More

Cumylamine 98.0+%, TCI America™

Catalog No. C12935ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12935ML 5 mL
C129325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12935ML Supplier TCI America Supplier No. C12935ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 585-32-0
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00134680
InChI Key KDFDOINBXBEOLZ-UHFFFAOYSA-N
Synonym cumylamine, alpha,alpha-dimethylbenzylamine, 1-methyl-1-phenyl-ethylamine, 1-methyl-1-phenylethylamine, 2-phenyl-2-propanamine, alpha-aminoisopropylbenzene, benzenemethanamine, alpha,alpha-dimethyl, dimethyl-aminophenylmethane, cumyl amine, phenisopropyl amine
PubChem CID 68509
IUPAC Name 2-phenylpropan-2-amine
SMILES CC(C)(N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 197°C
Quantity 5 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cumylamine
EINECSNumber (3)-0332&(3)-2693
TSCA No