missing translation for 'onlineSavingsMsg'
Learn More

Cumyl hydroperoxide, 80%

Catalog No. p-94486
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC349960050 5 g Plastic bottle
AC349962500 250 g Plastic bottle
AC349960010 1 kg Plastic bottle
AC349960025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC349960050 Supplier Thermo Scientific Chemicals Supplier No. 349960050
Only null left

CAS: 80-15-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 80-15-9 , 98-82-8 , 617-94-7 , 98-86-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
Synonym cumene hydroperoxide, cumyl hydroperoxide, hydroperoxide, 1-methyl-1-phenylethyl, cumenyl hydroperoxide, alpha,alpha-dimethylbenzyl hydroperoxide, cumolhydroperoxid, 7-cumyl hydroperoxide, cumolhydroperoxide, cument hydroperoxide, hydroperoxyde de cumene
PubChem CID 6629
ChEBI CHEBI:78673
SMILES CC(C)(OO)C1=CC=CC=C1

Specifications

Melting Point -30.0°C
Density 1.0600g/mL
Flash Point 83°C
Assay Percent Range 80 to 85% (Iodometry)
Packaging Plastic bottle
Linear Formula C6H5C(CH3)2OOH
Quantity 5 g
Specific Gravity 1.06
Solubility Information Solubility in water: miscible.
IUPAC Name 2-hydroperoxypropan-2-ylbenzene
Formula Weight 152.19
Percent Purity 80%
Chemical Name or Material Cumyl hydroperoxide, 80%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Heating may cause a fire.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause re
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
EINECSNumber 201-254-7
RTECSNumber MX2450000
TSCA TSCA

RUO – Research Use Only