missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis III Micro Balance

Catalog No. 14561545
Change view
Click to view available options
Capacity:
1.1 g, 2.1 g, 3.1 g
2.1 g
6.1 g
10.1 g
Includes:
Draft Shield A
Draft Shield A QP99
Draft Shield F
Draft Shield F QP99
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes
14561545 2.1 g Draft Shield A
14561513 1.1 g, 2.1 g, 3.1 g Draft Shield A
14561535 1.1 g, 2.1 g, 3.1 g Draft Shield A QP99
14561546 2.1 g Draft Shield A QP99
14561507 2.1 g Draft Shield F
14561452 2.1 g Draft Shield F QP99
14561519 6.1 g Draft Shield A
14561469 6.1 g Draft Shield A QP99
14561499 6.1 g Draft Shield F
14561537 6.1 g Draft Shield F QP99
14561502 10.1 g Draft Shield A
14561414 10.1 g Draft Shield A QP99
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14561545 Supplier Sartorius Supplier No. CUB2.7S1S2A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Electronic micro balance with 7 in. color TFT touch display.

  • Deliver precise, reliable weighing results for virtually any regulated lab application, with capacities from 2.1 g to 70 kg and readabilities from 0.1 μg to 1 g.
  • A flexible range of weighing modules, draft shields, and software options lets customers tailor the balances to their workflow, compliance, and integration needs.
  • Upgradeable hardware and software protect your investment and keep your solution ready for future requirements.
  • Built for digital labs, Cubis III enables secure, plug-and-play integration with LIMS, ELN, and other lab systems via standards such as OPC UA, LADS, and SiLA.
  • This helps simplify onboarding of new balances, accelerates validation, and reduces integration costs, while providing a scalable foundation for automation.
  • As connectivity becomes essential to GxP operations, Cubis III delivers real business value by strengthening data integrity, improving audit readiness, accelerating batch release, reducing deviations, and lowering costs.
  • Fits seamlessly into connected labs for a more efficient, integrated, future-ready workflow.

Specifications

Capacity 2.1 g
Display Type Color TFT Touchscreen
Readability 0.0001 mg
Includes Draft Shield A
Type Micro Balance