missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Ultra-High Resolution Semi-Micro Balances MCA
GREENER_CHOICE

Catalog No. p-200087472
Change view
Click to view available options
Model:
HWL Configuration
ION Configuration
MDS Configuration
QP99 Configuration
QP99 HWL Configuration
QP99 ION Configuration
QP99 MDS Configuration
Standard Configuration
Capacity:
220 g, 120 g
120 g
220 g
Readability:
0.1 mg, 0.01 mg
0.005 mg
0.01 mg
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Capacity Readability
14996856 Standard Configuration 220 g 0.005 mg
14996855 HWL Configuration 220 g 0.005 mg
14996854 ION Configuration 220 g 0.005 mg
14996851 QP99 ION Configuration 220 g 0.005 mg
14996853 MDS Configuration 220 g 0.005 mg
14996850 QP99 MDS Configuration 220 g 0.005 mg
14996852 QP99 Configuration 220 g 0.005 mg
14996875 Standard Configuration 120 g 0.01 mg
14996874 HWL Configuration 120 g 0.01 mg
14996873 ION Configuration 120 g 0.01 mg
14996872 MDS Configuration 120 g 0.01 mg
14996863 Standard Configuration 220 g 0.01 mg
14996862 HWL Configuration 220 g 0.01 mg
14996861 ION Configuration 220 g 0.01 mg
14996858 QP99 ION Configuration 220 g 0.01 mg
14996860 MDS Configuration 220 g 0.01 mg
14996857 QP99 MDS Configuration 220 g 0.01 mg
14996859 QP99 Configuration 220 g 0.01 mg
14996871 Standard Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996870 HWL Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996866 QP99 HWL Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996869 ION Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996865 QP99 ION Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996868 MDS Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996864 QP99 MDS Configuration 220 g, 120 g 0.1 mg, 0.01 mg
14996867 QP99 Configuration 220 g, 120 g 0.1 mg, 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 14996856 Supplier Sartorius Supplier No. MCA226S3S00D

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Cubis™ II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications, and hardware functions.

Superior weighing performance

Fast measurement time and accurate weighing results are guaranteed thanks to the next generation monolithic weighing system and the engineering solution compen- sating for environmental changes.

No more issues with charged samples

The glass draft shields coated with conductive layer prevents from outer electrostatic effects and the built-in ionizer effectively eliminates electrostatic charges from samples.

Cleaning process guidance

Intuitive process for daily and advanced cleaning is now available in all Cubis™ II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as tracking of these events.

Hardware upgradeability

Hardware features like motorized draft shield function or built-in ionizer can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended to use under challenging environmental conditions (e.g. in drafty environment).

Compliance and Data Integrity

The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis™ II balances, without the need of additional software.

Balances Fleet Management

The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis™ II MCA lab balance fleet across all labs within the same network.

Ease of use

The teaching function with learning capability of the motorized draft shield, guided workflows for many different applications (QApps), automated motorized leveling and automatic internal adjustment (isoCAL) help to with easy and error-free operation of the balance.

Specifications

Capacity 220 g
Display Type Color TFT Touchscreen
Readability 0.005 mg
Model Standard Configuration
Type Semi-micro Balance