Learn More
Sartorius Cubis II MTS Balance PROMO
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.
$11330.77 - $16243.25
Specifications
| Connection Type | D-sub connector, 9-pin |
|---|---|
| Display Type | Color Touchscreen |
| Type | Balance |
| Catalog Number | Mfr. No. | Model | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Price | Quantity | |||||
|
01288035PM
|
Sartorius
MCA8202S0MTSPM |
MCA8202S0MTSPM |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
01288034PM
|
Sartorius
MCE8202S0MTSPM |
MCE8202S0MTSPM |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Leading performance.
- Monolithical weighing system, integrated climate sensors, individual sample holders
- Error-free operation.
- Individual QApp workflows, motorized auto-leveling
- The modular weighing system Cubis™ II can be individually configured from different components.
Specifications
| D-sub connector, 9-pin | |
| Color Touchscreen | |
| Balance |