missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis II MTS Balance PROMO

The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.

Supplier:  Sartorius MCE8202S0MTSPM

 View more versions of this product

Catalog No. 01288034PM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Leading performance.
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling
  • The modular weighing system Cubis™ II can be individually configured from different components.
Specifications

Specifications

D-sub connector, 9-pin
8200 g
10 mg
MCE8202S0MTSPM
Balance
MCE Display with factory-installed essential weighing applications
Color Touchscreen
Die-cast Aluminum, Plastic, Optiwhite Float Glass and Stainless Steel
1 sec.
Class II
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions