missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CU-T12-9

Catalog No. 541410
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
541410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 541410 Supplier Tocris Bioscience™ Supplier No. 5414/10
Only null left

Potent TLR1/2 agonist

CU-T12-9 is a potent TLR1/2 agonist (EC50 = 52.9 nM). Activates the NF-κB pathway and upregulates proinflammatory cytokines in vitro. Also enhances TLR1 and TLR2 dimerization.

Specifications

Chemical Name or Material CU-T12-9
CAS 1821387-73-8
Synonym N-Methyl-4-nitro-2-(4-(4-trifluoromethyl)phenyl)-1H-imidazol-1-yl)aniline
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C17H13F3N4O2
Formula Weight 362.31
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use