missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ DPBS, calcium, magnesium

Catalog No. A1285801
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1285801 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1285801 Supplier Gibco™ Supplier No. A1285801
Only null left

Gibco Cell Therapy Systems DPBS CTS™ is the classical Dulbecco's Phosphate Buffered Saline which contains calcium, magnesium but no phenol red.

Gibco Cell Therapy Systems DPBS CTS is the classical Dulbecco's Phosphate Buffered Saline which contains calcium, magnesium but no phenol red.

Gibco DPBS CTS is manufactured in state-of-the-art cGMP, ISO certified facilities to help ensure the highest quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, the absence of bacterial and fungal contamination and endotoxin tests.

Gibco CTS products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

This product will have the following intended use specific for cell therapy market - For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals

Specifications

Chemical Name or Material DPBS
Concentration 1X
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Physical Form Liquid
Quantity 1000 mL
Solution Type Dulbecco's Phosphate Buffered Saline
Packaging Bottle
Sterility Sterile-filtered
Includes Calcium and Magnesium
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21 CFR820

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.