Learn More
Gibco™ CTS™ ACK Lysing Buffer
CTS ACK (Ammonium-Chloride-Potassium) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow.
$48.11 - $48.11
Specifications
Form Clear, colorless solution
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Format Price Quantity  
Catalog Number Mfr. No. Quantity Format Price Quantity  
A4000225501
View Documents
Gibco™
A4000225501
100 mL Bottle
Each for $48.11
Only null left
Description
Description
Gibco Cell Therapy Systems (CTS) ACK (Ammonium-Chloride-Potassium) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow. The complete formulation and lysis protocol are available.
CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.
Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.
Specifications
Specifications
Clear, colorless solution
Videos
SDS
Documents
Documents
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.