missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AAV-MAX Lysis Buffer

Catalog No. a5152103
Change view
Click to view available options
Quantity:
100 mL
1 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5152103 10 L
A5152101 100 mL
A5152102 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5152103 Supplier Gibco™ Supplier No. A5152103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco CTS AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells.

Gibco CTS AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells. The cell lysis is detergent-based, eliminating the need for mechanical cell disruption, providing a milder and easier alternative when isolating AAV particles from cell cultures. The buffer is supplied as a 10X solution that can be directly added to HEK293 AAV production cultures to induce cell lysis.

Note: the formulation of this product differs from the formulation of our research-use-only AAV-MAX Lysis Buffer (Cat. No. A50520). This formulation reduces sample turbidity and precipitation formation post-clarification, including when samples are stored between clarification and subsequent downstream workflow steps.

Features of CTS AAV-MAX Lysis Buffer include:
• Polysorbate 20-based formulation that does not contain Triton X-100
• Formulated with 20 mM MgCl2 for convenience
• Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File Letter of Authorization and Regulatory Support File available upon request.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and CTS versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (Cat. No. A48400), CTS AAV-MAX Transfection Kit (A5427701), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

Specifications

Concentration 10X
Product Type Cell Lysis Buffer
Content And Storage Store at 2–8°C.

Form Liquid
Product Line AAV-MAX
Reagent Type Cell Lysis Buffer
Quantity 10 L
pH 6–8

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.