missing translation for 'onlineSavingsMsg'
Learn More

CTI SGPF Series Sterile Latex Cleanroom Gloves

Sterile, powder free latex gloves are qualified for use in the pharmaceutical industry and recommended for Class 100 /ISO 5 cleanroom compatible or higher controlled environments.

$529.01 - $566.18

Specifications

Certifications/Compliance ASTM D3578-95, IEST-RP-CC005.2, FDA 21 CFR 177.2600, AAMI, MIL STD-105E
Cleanroom Class Class 100 (ISO 5)
Color White
Exterior Finish Fully Textured
Hand Specific
View More Specs

Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-999-102A
View Documents
CTI
SGPF600
6
Case of 200 Each for $529.01
Only null left
 
18-999-102B
View Documents
CTI
SGPF650
6.5
Case of 200 Pair for $529.01
Only null left
 
18-999-102C
View Documents
CTI
SGPF700
7
Case of 200 Pair for $529.01
Only null left
 
18-999-102D
View Documents
CTI
SGPF750
7.5
Case of 200 Pair for $529.01
Only null left
 
18-999-102E
View Documents
CTI
SGPF800
8
Case of 200 Pair for $529.01
Only null left
 
18-999-102F
View Documents
CTI
SGPF850
8.5
Case of 200 Pair for $529.01
Only null left
 
18-999-102G
View Documents
CTI
SGPF900
9
Case of 200 Pair for $566.18
Only null left
 
18-999-102H
View Documents
CTI
SGPF1000
10
Case of 200 Pair for $566.18
Only null left
 
Description

Description

  • Constructed of double chlorinated latex
  • Low protein and contact sensitizers
  • Textured finish for secure grip
  • Tapered, beaded cuff offers easy donning and doffing
  • Hand-specific
  • Thickness: 6.5 mil
  • Length: 12 in. (30.5 cm)
  • Color: White

Specifications

Specifications

ASTM D3578-95, IEST-RP-CC005.2, FDA 21 CFR 177.2600, AAMI, MIL STD-105E
White
Specific
Powder Free
6.5 mil
Yes
SGPF Series
12 in.
Class 100 (ISO 5)
Fully Textured
Latex
Sterile
0.16 mm
200 Pairs/Case (4 Boxes of 50 Pks of Gloves, Each Pk Contains 1 Pair)
23 MPa
30.5 cm
Videos
SDS
Documents

Documents