missing translation for 'onlineSavingsMsg'
Learn More

Crystal Violet Indicator, 1% (w/v) in Glacial Acetic Acid, Ricca Chemical

Gentian Violet Indicator for Nonaqueous Titrations

$188.64 - $309.03

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) Mixture
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2483-4
SDS View Documents
Ricca Chemical Company
24834
120 mL Amber Glass Bottle
Each for $188.64
Only null left
 
2483-16
SDS View Documents
Ricca Chemical Company
248316
500 mL Amber Glass Bottle
Each for $309.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

548-62-9
Mixture
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

548-62-9 , 64-19-7
C25H30ClN3
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Mixture
CHEBI:41688
Indicator
Glacial Acetic Acid
<2
Liquid
1% (w/v)
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
1% W/V
Amber Glass Bottle
Blue to Purple/Violet
120 mL
Crystal Violet (Gentian Violet) Indicator
Videos
SDS
Documents

Documents