missing translation for 'onlineSavingsMsg'
Learn More

Crotyl Bromide 83.0+%, TCI America™

Catalog No. B056225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B056225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B056225ML Supplier TCI America Supplier No. B056225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4784-77-4
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00000248
InChI Key AVMHMVJVHYGDOO-IHWYPQMZSA-N
Synonym 1-Bromo-2-butene
PubChem CID 5354843
IUPAC Name (2Z)-1-bromobut-2-ene
SMILES C\C=C/CBr

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 mL
UN Number 1993
Formula Weight 135.00
Percent Purity ≥83.0% (GC)
Physical Form Liquid
Chemical Name or Material Crotyl Bromide
TSCA Yes
Recommended Storage Refrigerator