accessibility menu, dialog, popup

UserName

CH3Br(Z)

CAS RN 4784-77-4

IUPAC Name: (2Z)-1-bromobut-2-ene
Synonyms: (2Z)-1-bromobut-2-ene
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: AVMHMVJVHYGDOO-IHWYPQMZSA-N
SMILES: C\C=C/CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results