missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPLX1 Monoclonal Antibody (K40066_9A4), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | CPLX1 |
|---|---|
| Clone | K40066_9A4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: GATKDMGKML GGDEEKDPDA AKKEEERQEA LRQAEEERKA KYAKMEAERE AVRQGIRDKY GIKKKEEREA EAQAAMEANS EGSLTRPKKA IPPGCGDEVE EEDESILDTV
CPLX1 Monoclonal antibody specifically detects CPLX1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CPLX1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| 921-S; complexin 1; complexin I; Complexin-1; Cplx1; CPX I; CPX1; CPX-I; synaphin 2; synaphin-2 | |
| Recombinant human CPLX1 (immunogen range is 11-120). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Cplx1 |
| K40066_9A4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| O14810, P63040, P63041 | |
| 10815, 12889, 64832 | |
| IgG1 | |
| Protein A | |
| Antibody |