missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CPLX1 Monoclonal Antibody (K40066_9A4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556004
Description
Sequence of this protein is as follows: GATKDMGKML GGDEEKDPDA AKKEEERQEA LRQAEEERKA KYAKMEAERE AVRQGIRDKY GIKKKEEREA EAQAAMEANS EGSLTRPKKA IPPGCGDEVE EEDESILDTV
CPLX1 encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release.Specifications
| CPLX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| O14810, P63040, P63041 | |
| Cplx1 | |
| Recombinant human CPLX1 (immunogen range is 11-120). | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K40066_9A4 | |
| Unconjugated | |
| Cplx1 | |
| 921-S; complexin 1; complexin I; Complexin-1; Cplx1; CPX I; CPX1; CPX-I; synaphin 2; synaphin-2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10815, 12889, 64832 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |