missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CpG oligodeoxynucleotides Mouse

Catalog No. NBP231141SS
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231141SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231141SS Supplier R&D Systems™ Supplier No. NBP231141SS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 0.025 mg
For Use With (Application) Activation of mouse TLR9
Type CpG Oligodeoxynucleotides