missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CpG oligodeoxynucleotides Mouse

Catalog No. NBP231139SS
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231139SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231139SS Supplier Novus Biologicals™ Supplier No. NBP231139SS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.025 mg
For Use With (Application) Activation of mouse TLR9
Type CpG Oligodeoxynucleotides