missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CpG oligodeoxynucleotides Mouse

Catalog No. NBP231132
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231132 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231132 Supplier Novus Biologicals™ Supplier No. NBP231132

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Applications: Ligand Activation

Specifications

Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1 mg
For Use With (Application) Activation of mouse TLR9
Type CpG Oligodeoxynucleotides