missing translation for 'onlineSavingsMsg'
Learn More

CpG oligodeoxynucleotides (2216), Novus Biologicals™

Catalog No. NBP231136
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231136 0.1 mg
NBP231136SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231136 Supplier Bio-Techne Supplier No. NBP231136

Useful positive control model system for studying hTLR9 activation

Immunogen: G&asterisk;GGGGACGATCGTCG&asterisk;G&asterisk;G&asterisk;G&asterisk;G&asterisk;G Buffer: Sterile water Clone: 2216 Dilution: Ligand Activation, 5 to 20μg/mL CpG ODN Type A (2216)

Specifications

Concentration 1.0 mg/mL
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1 mg
For Use With (Application) Human CpG ODNs are used for activation of human TLR9 at a range of 5 to 20μL. The TLR9/NF-kB/SEAPorter HEK 293 cell line is a useful positive control model system for studying hTLR9 activation. Typically use a 96-well plate format assay using this reporter cell line.
Type CpG Oligodeoxynucleotides
Species Human