Learn More
Corning™ Costar™ Spin-X™ Centrifuge Tube Filters, CA Membrane
Consisting of a membrane-containing filter unit within a polypropylene centrifuge tube. Corning™ Costar™ Spin-X™Centrifuge Tube Filters effectively filters small sample volumes by centrifugation for bacteria removal, particle removal, HPLC sample preparation, removal of cells from media and purification of DNA from agarose and polyacrylamide gels.
Supplier:  Corning™ 8161
 View more versions of this product
Catalog No. 07200386
Only null left
Description
Description
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Specifications
Chemical Identifiers
Cellulose Acetate
Spin-X
0.5 mL
Specifications
Cellulose Acetate
Non-sterile
0.5 mL
0.22 μm
Spin-X
Product Suggestions
Product Suggestions