Learn More
Pyramex Safety Products GL604C5 Series Sandy Nitrile-Coated HPPE Gloves
Sandy nitrile coating provides enhanced grip capabilities in dry, wet and oily conditions.
$170.27 - $170.27
Specifications
ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420
Coating Material Nitrile
Color Gray, Black
Cut Resistance Rating ANSI A4
View More Specs
Products 5
Catalog Number Mfr. No. Size Glove Cuff Color Price Quantity  
Catalog Number Mfr. No. Size Glove Cuff Color Price Quantity  
19555079
View Documents
Pyramex Safety Products
GL604C5S
Small Red
Pack of 12 for $170.27
Only null left
19555078
View Documents
Pyramex Safety Products
GL604C5M
Medium Yellow
Pack of 12 for $170.27
Only null left
19555077
View Documents
Pyramex Safety Products
GL604C5L
Large Brown
Pack of 12 for $170.27
Only null left
19555081
View Documents
Pyramex Safety Products
GL604C5XL
X-Large Black
Pack of 12 for $170.27
Only null left
19555080
View Documents
Pyramex Safety Products
GL604C5X2
2X-Large Blue
Pack of 12 for $170.27
Only null left
Description
Description
  • Sandy nitrile coated gloves
  • 13-gauge HPPE shell is resistant to tears and abrasion
  • Enhances grip capabilities in dry, wet, and oily conditions
Specifications
Specifications
ANSI 6
Nitrile
ANSI A4
No
Cut-resistant Gloves
Assembly lines, manufacturing, HVAC, construction
Sandy palm
Sandy Nitrile GL604C5 Series
EN 420
Gray, Black
Specific
HPPE
EN 388:4X43D
Elasticated
ANSI/ISEA 105-16 Level 3
13
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
WARNING: Cancer and Reproductive Harm - www.P65warnings.ca.gov