missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cortisone, 98%

Supplier:  Thermo Scientific™ 460520010

Quantity 1g

 View more versions of this product

Catalog No. AC460520010


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

53-06-5
360.45
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
222786
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
C21H28O5
MFCD00003610
cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
CHEBI:16962
C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Cortisone
2% max.
97.5% min. (HPLC)
C21H28O5
1g
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
(8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione
222786
360.44
53-06-5
Authentic
Glass Bottle
MFCD00003610
cortisone, andreson, cortisal, cortisate, cortistal, cortivite, cortogen, cortone, kendall's compound e, adrenalex
C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
360.45
CHEBI:16962
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging the unborn child.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 200-162-4

SDS
Documents

Documents

Promotions

Promotions