CAS RN 53-06-5
IUPAC Name:
(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
Synonyms:
cortisone therapeutic cortisone cortison cortivite kendall's compound kendall compound E cortisonum cortisone preparation reichstein fa 可的松
Molecular Weight (g/mol):
360.45
Molecular Formula:
C21H28O5
InChi Key:
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
SMILES:
C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific™ Cortisone, 98%
Thermo Scientific™ Cortisone, 98%
CAS: 53-06-5 Molecular Formula: C21H28O5 Molecular Weight (g/mol): 360.45 MDL Number: MFCD00003610 InChI Key: MFYSYFVPBJMHGN-ZPOLXVRWSA-N Synonym: cortisone,andreson,cortisal,cortisate,cortistal,cortivite,cortogen,cortone,kendall's compound e,adrenalex PubChem CID: 222786 ChEBI: CHEBI:16962 IUPAC Name: (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthrene-3,11-dione SMILES: C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO