Learn More
Corning™ Single-Use Bags for HYPERStack™ Vessels
Used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution
$436.73 - $477.94
Specifications
Film Description ULDPE
No. per Pack 1/Pk.
Quantity Each
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Tubing Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Tubing Style Price Quantity & Availability  
MT9120077
View Documents Promotion Details
Corning™
9120077
20 L Heat sealable flexible tubing
Each for $436.73
Only null left
MT9120075
View Documents Promotion Details
Corning™
9120075
5 L Heat sealable flexible tubing
Each for $450.09
Only null left
MT9120076
View Documents Promotion Details
Corning™
9120076
5 L PVC tubing
Each for $477.94
Only null left
Description
Description
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors. They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Film type: ULDPE
Corning media certificates are available on our Resource Library.
Specifications
Specifications
ULDPE
1/Pk.
Each