Learn More
Corning™ Single-Use Bags for HYPERStack™ Vessels
Used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution
Supplier:  Corning™ 9120076
 View more versions of this product
Catalog No. MT9120076
Only null left
Description
Description
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors. They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Film type: ULDPE
Corning media certificates are available on our Resource Library.
Specifications
Chemical Identifiers
ULDPE
1/Pk.
Each
Specifications
Hyperstack™ vessels to add quenching substrates to complete cell culture in an entirely closed solution
ULDPE
1/Pk.
Each
PVC tubing
1/Cs.
14 L x 0.37 I.D. x 0.62 in. O.D. chemically resistant, heat sealable flexible tubing, female MPC connector with 0.37 in. barb male MPC cap, male luer connector with female luer cap, swabbable valve, bag port with 0.5 in. barb
5 L
Product Suggestions
Product Suggestions