Learn More
Corning™ Single-Use Bags for HYPERStack™ Vessels
Used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution
Supplier: Corning™ 9120075
Description
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors. They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.Film type: ULDPE
Corning media certificates are available on our Resource Library.
Chemical Identifiers
| ULDPE | |
| 1/Pk. | |
| Each |
Specifications
| Hyperstack™ vessels to add trypsin substrates to complete cell culture in an entirely closed solution | |
| ULDPE | |
| 1/Pk. | |
| Each |
| Heat sealable flexible tubing | |
| 1/Ea. | |
| Port 1-12 L x 0.118 I.D. x 0.164 in. O.D. PVC tubing, 0.37 in. x 0.12 in. reducer, Port 2-male MPC connector with 0.37 in. barb female MPC cap, male luer connector with female luer cap, swabbable valve, bag port with 0.5 in. barb | |
| 5 L |