missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Premium Fetal Bovine Serum
GREENER_CHOICE

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

$1,221.09 - $2,170.27

Specifications

Certifications/Compliance cGMP
Color Caramel/Amber
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Heat Inactivated Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Heat Inactivated Price Quantity & Availability  
MT35016CV
GREENER_CHOICE
View Documents Promotion Details
Corning™
35016CV
Fetal Bovine Serum Yes
Each for $2,170.27
Only null left
 
MT35015CV
GREENER_CHOICE
View Documents Promotion Details
Corning™
35015CV
Fetal Bovine Serum No
Each for $1,221.09
Only null left
 
Description

Description

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds

Compliance

USDA Certified FBS is sourced from USDA-certified facilities (abbatoirs).

Specifications

Specifications

cGMP
-25° to -15°C
500 mL
Triple-filtered, 0.1 μm
Premium
Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
7.8 ±0.8
USDA Inspected
Caramel/Amber
Fetal Bovine Serum
Sterile
Bovine Fetus
≤20EU/ mL
<30 mg/dL
30 to 50g/L
Videos
SDS
Documents

Documents