missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Premium Fetal Bovine Serum
GREENER_CHOICE

Catalog No. MT35015CV
Change view
Click to view available options
Product Type:
Fetal Bovine Serum
Heat Inactivated:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Heat Inactivated
MT35015CV Fetal Bovine Serum No
MT35016CV Fetal Bovine Serum Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT35015CV Supplier Corning™ Supplier No. 35015CV
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds

Compliance

USDA Certified FBS is sourced from USDA-certified facilities (abbatoirs).

Specifications

Certifications/Compliance cGMP
Color Caramel/Amber
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Premium
Endotoxin Concentration ≤20EU/ mL
For Use With (Application) Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
Heat Inactivated No
Hemoglobin Concentration <30 mg/dL
Osmolality 315 ± 35mOsm/kg
pH 7.8 ±0.8
Protein Content 30 to 50g/L
USDA Safety Testing Included USDA Inspected
Show More Show Less