Learn More
Corning™ PF Media
Optimized for the cultivation of derivatives of the parental Chinese Hamster Ovary cell line, specifically CHO K-1
Supplier:  Corning™ 40200CV
Catalog No. MT40200CV
Only null left
Explore available promotions
Description
Description
  • Proprietary serum-free and protein-free growth medium that does not contain any hormones or growth factors
  • Supports superior growth and viability over long-term pasasges in both adherent and suspension cell culture
  • Formulated without L-glutamine
For research use only. Not for IVD use.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
CHO
No Glutamine
6 x 500 mL
Product Suggestions
Product Suggestions