Learn More
Corning™ Trace Elements
For Research applications
Specifications
Form Liquid
Product Type Trace Element
Sterility Sterile-filtered
Format Plastic Bottle
Quantity 100 mL
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
MT99182CI
View Documents
Corning™
25021CI
Trace Elements A N/A
Only null left
MT99176CI
View Documents
Corning™
25023CI
Trace Elements C N/A
Only null left
MT99175CI
View Documents
Corning™
25022CI
Trace Elements B N/A
Only null left
Description
Description
cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride
Specifications
Specifications
Liquid
Sterile-filtered
100 mL
Trace Element
Plastic Bottle