missing translation for 'onlineSavingsMsg'
Learn More

Corning™ L-glutamine Powder

An essential amino acid and a key component of culture media

$309.83 - $309.83

Specifications

Content And Storage 2° to 8°C
Form Powder
Product Type L-Glutamine
Format Plastic Bottle
pH 5 to 6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT61030RO
View Documents
Corning™
61030RO
500 g
Each for $309.83
Only null left
 
Description

Description

L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use. This formulation is prepared in cell culture grade water.

  • Complete Certificate of Analysis available for each production lot, along with full formulation information
  • Produced under the highest industry standards to ensure superior results
Specifications

Specifications

2° to 8°C
L-Glutamine
5 to 6
Powder
Plastic Bottle
Videos
SDS
Documents

Documents