missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cell Culture Tris-Buffered Saline (10X), Corning™
Description
- Maintains cell culture media in the physiological pH range
- Maintains cell tonicity and viability
- Produced under the highest industry standards to ensure superior results
- Nontoxic
Specifications
Specifications
| CAS | 77-86-1, 7732-18-5 |
| Concentration | 10X |
| pH | 7.4 |
| Physical Form | Liquid |
| Quantity | 6 x 1000 mL |
| Solution Type | Wash Buffer |
| Formulation | Sodium Chloride 80g/L, Tris Ultrapure 24.2g/L |
| Grade | Molecular Biology |