missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X

Catalog No. p-4903138
Change view
Click to view available options
Quantity:
1 x 10,000 mL
1 x 20,000 mL
1 x 50,000 mL
6 x 500 mL
6 x 1000 mL
Physical Form:
Liquid
Powder
Includes:
Calcium and Magnesium
No Calcium, No Magnesium
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Physical Form Includes
MT21030CM 6 x 1000 mL Liquid Calcium and Magnesium
MT21031LB 1 x 20,000 mL Liquid No Calcium, No Magnesium
MT21031LX 1 x 10,000 mL Liquid No Calcium, No Magnesium
MT55031PB 1 x 10,000 mL Powder No Calcium, No Magnesium
MT55031PC 1 x 50,000 mL Powder No Calcium, No Magnesium
MT21030CV 6 x 500 mL Liquid Calcium and Magnesium
MT21031CM 6 x 1000 mL Liquid No Calcium, No Magnesium
MT21031CV 6 x 500 mL Liquid No Calcium, No Magnesium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MT21030CM Supplier Corning™ Supplier No. 21030CM
Only null left

With low endotoxin content.

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. This formulation contains calcium and magnesium.

Specifications

Chemical Name or Material DPBS
Concentration 1 X
pH 6.9 to 7.1
Physical Form Liquid
Quantity 6 x 1000 mL
Formulation Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L
Includes Calcium and Magnesium