missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 10% SDS (Sodium Dodecyl Sulfate)

Powerful protein denaturant produced under highest industry standards

Supplier:  Corning™ 46040CI

Catalog No. MT46040CI


Only null left
Explore available promotions

Description

Description

  • Anionic wetting agent for both acid and alkaline solutions
  • For protein and lipid solubilization in SDS-PAGE protein analysis and nucleic acid purification
  • Prepared in 18 megaohm water and is 0.2μm filtered

Specifications

Specifications

15° to 30°C
Liquid
10% solution is prepared in 18megaohm Water and is 0.2μm filtered
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions