missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
$302.25 - $908.44
Specifications
Closure Color | Clear |
---|---|
Closure Included | Closure Included |
Closure Material | Polystyrene |
Bottom Shape | Round |
Color | Clear |
Catalog Number | Mfr. No. | Diameter (Metric) | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Quantity | Price | Quantity | ||||
08-774-257
|
Corning™
354467 |
35 mm | 20/cs. |
Case of 20 Each for $324.16
|
|
||||
08-774-258
|
Corning™
356467 |
35 mm | 100/cs. |
Case of 100 Each for $815.19
|
|
||||
08-774-259
|
Corning™
354468 |
60 mm | 20/cs. |
Case of 20 Each for $302.25
|
|
||||
08-774-261
|
Corning™
354469 |
100 mm | 10/cs. |
Case of 10 Each for $306.74
|
|
||||
08-774-260
|
Corning™
356468 |
60 mm | 100/cs. |
Case of 100 Each for $908.44
|
|
||||
08-774-262
|
Corning™
356469 |
100 mm | 40/cs. |
Case of 40 Each for $738.18
|
|
||||
08-774-281
|
Corning™
354550 |
150 mm | 5/cs. |
Case of 5 Each for $470.90
|
|
||||
Description
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
- Coated with 0.2 micron filtered PDL in a cleanroom
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Round | |
Cell Culture Dish |
Closure Included | |
Round | |
Polystyrene | |
Poly-D-Lysine | |
Lid |