Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Supplier: Corning™ 354550
Description
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
- Coated with 0.2 micron filtered PDL in a cleanroom
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
| Clear | |
| Polystyrene | |
| Clear | |
| Gridded | |
| Round | |
| Cell Culture Dish | |
| 5/cs. |
| Closure Included | |
| Round | |
| 151.9 sq. cm | |
| Polystyrene | |
| Poly-D-Lysine | |
| Lid | |
| 150 mm |