missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium, Magnesium or Sodium Bicarbonate

Used to enzymatically release adherent cells from tissue culture plates for passaging.

$96.15 - $358.84

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium, No Sodium Bicarbonate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT25052CV
View Documents
Corning™
25052CV
6 x 500 mL
Case of 6 Each for $358.84
Only null left
 
MT25052CI
View Documents
Corning™
25052CI
6 x 100 mL
Case of 6 Each for $96.15
Only null left
 
Description

Description

Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging.  Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action.  EDTA sequesters these ions and thus enhances the efficacy of trypsin.  This product contains trypsin at a concentration of 0.05% with 0.53 mM EDTA, and does not contain sodium bicarbonate.
Specifications

Specifications

-20°C
Cell Culture Dissociation Reagent
No Calcium, No Magnesium, No Sodium Bicarbonate
Liquid
Sterile
7 to 7.6
Videos
SDS
Documents

Documents