missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, 1.053 Specific Gravity at 25°C (77°F), Ricca Chemical
For Hemoglobin Estimation in Blood Donor Screening of Men and Women
$123.69 - $123.69
Chemical Identifiers
| CAS | 7758-99-8 |
|---|---|
| Molecular Formula | CuH10O9S |
| Molecular Weight (g/mol) | 249.68 |
| MDL Number | MFCD00149681 |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
| PubChem CID | 24463 |
| ChEBI | CHEBI:31440 |
| IUPAC Name | copper(2+) pentahydrate sulfate |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Chemical Identifiers
| 7758-99-8 | |
| 249.68 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| 24463 | |
| copper(2+) pentahydrate sulfate |
| CuH10O9S | |
| MFCD00149681 | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| CHEBI:31440 | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Specifications
| 7758-99-8,7732-18-5 | |
| 94.0,7.83 | |
| MFCD00149681 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| copper(2+) pentahydrate sulfate | |
| 24463 | |
| Laboratory | |
| ∼4 | |
| Copper Sulfate Solution, Specific Gravity 1.053 |
| 90.0,7.52 | |
| CuH10O9S | |
| copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
| 249.68 | |
| CHEBI:31440 | |
| Amber Glass Bottle | |
| 500 mL |