missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate Solution (For AOAC Method 930.28), Fisher Chemical™

Catalog No. SC9202250
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SC9202250 250 mL Natural Poly Bottle
SC9202500 500 mL Natural Poly Bottle
SC92021 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SC9202250 Supplier Ricca Chemical Company Supplier No. SC9202250

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

CAS Min % 92.0, 6.35
CAS Max % 95.0, 6.61
Grade Laboratory
Packaging Natural Poly Bottle
Methods For AOAC Method 930.28
Quantity 250 mL
Chemical Name or Material Copper Sulfate Solution