missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate Solution, 1.6% (w/v) in 0.4% (v/v) Sulfuric Acid, Ricca Chemical

Catalog No. R23186001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R23186001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R23186001A Supplier Ricca Chemical Company Supplier No. R23186001A
Only null left

Chemical Identifiers

CAS 7664-93-9 , 7758-99-8 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Assay Percent Range 1.60 ± 0.02% (w/v)
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH <2
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Copper Sulfate Solution, 1.6% (w/v) in 0.4% (v/v) Sulfuric Acid
Show More Show Less