missing translation for 'onlineSavingsMsg'
Learn More

Copper Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCU22Y

 View more versions of this product

Catalog No. PLCU2-2Y


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cu1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Copper Standard
7440-50-8,7697-37-2,7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,2.0,97.9
MFCD00010965
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
RYGMFSIKBFXOCR-UHFFFAOYSA-N
2% HNO3
63.55
23978
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
0.1,2.0,97.9
Cu
AA, ICP
Certified Reference Material
[Cu]
copper
125 mL
CHEBI:30052

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
CHEBI:30052
[Cu]
Cu
MFCD00010965
23978
copper
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only