missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$255.35 - $510.68
Chemical Identifiers
| CAS | 7440-50-8 |
|---|---|
| Molecular Formula | Cu |
| Molecular Weight (g/mol) | 63.55 |
| MDL Number | MFCD00010965 |
| InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
| PubChem CID | 23978 |
| ChEBI | CHEBI:30052 |
| IUPAC Name | copper |
| SMILES | [Cu] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLCU2-2Y
|
SPEX CertiPrep Group LLC
PLCU22Y |
125 mL | Triple-leached LDPE bottle |
Each for $255.35
|
|
||||
|
PLCU22T
|
SPEX CertiPrep Group LLC
PLCU22T |
250 mL | Triple-leached LDPE bottle |
Each for $339.59
|
|
||||
|
PLCU2-2X
|
SPEX CertiPrep Group LLC
PLCU22X |
500 mL | Triple-leached LDPE bottle |
Each for $510.68
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Cu | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-50-8 | |
| 63.55 | |
| RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
| CHEBI:30052 | |
| [Cu] |
| Cu | |
| MFCD00010965 | |
| 23978 | |
| copper |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Cu | |
| AA, ICP | |
| Certified Reference Material | |
| [Cu] | |
| copper | |
| 125 mL | |
| CHEBI:30052 | |
| Copper Standard |
| 7440-50-8,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00010965 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 63.55 | |
| 23978 | |
| Assurance |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only