missing translation for 'onlineSavingsMsg'
Learn More

Copper phthalocyanine, dye content ca 95%

CAS: 147-14-8 | C32H16CuN8

$89.90 - $501.10

Chemical Identifiers

CAS 147-14-8
Molecular Formula C32H16CuN8
MDL Number MFCD00010719
Synonym C.I. 74160, Pigment blue 15
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC269150250
View Documents Promotion Details
N/A 25 g Glass bottle
Each for $89.90
Only null left
 
AC269152500
View Documents Promotion Details
N/A 250 g Glass bottle
Each for $501.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

147-14-8
C32H16CuN8
MFCD00010719
C.I. 74160, Pigment blue 15

Specifications

147-14-8
MFCD00010719
C.I. 74160, Pigment blue 15
Authentic
≥94% (ex Cu)
Blue
Copper phthalocyanine, dye content ca 95%
C32H16CuN8
15, 2505
Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons
576.08
Complies
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-685-1

RTECSNumber : GL8510000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only