missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, anhydrous, 98%

Copper(II) sulfate, 98%, CuO4S, CAS Number-7758-98-7, copper 2+ sulfate, copper ii sulfate, blue stone, copper monosulfate, copper sulphate, trinagle, copper sulfate, hylinec, cupric sulfate anhydrous, copper ii sulfate, anhydrous, 1000g, CHEBI:23414, 3.6, Hazard Class: 9; Packaging Group: III | CAS: 7758-98-7 | CuO4S | 159.602 g/mol

Supplier:  Thermo Scientific Chemicals A139860B

 View more versions of this product

Catalog No. AAA139860B


Only null left
Explore available promotions

Description

Description

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong bases, metals, alkali metals and powdered metals.
Specifications

Specifications

Copper(II) sulfate
anhydrous
1000 g
CuSO4
UN3288
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper;sulfate
24462
159.6
Odorless
3.6
>560°C
7758-98-7
CuO4S
MFCD00010981
14,2653
Soluble in water and methanol. Insoluble in ethanol.
[O-]S(=O)(=O)[O-].[Cu+2]
159.602
CHEBI:23414
98%
Hygroscopic

Chemical Identifiers

7758-98-7
159.602
ARUVKPQLZAKDPS-UHFFFAOYSA-L
24462
copper;sulfate
CuO4S
MFCD00010981
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
CHEBI:23414
[O-]S(=O)(=O)[O-].[Cu+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-847-6

RTECSNumber : GL8800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only